Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Difluoro-3-methylphenol, 97+%

Click to view available options
Quantity:
1g
5g
261763-46-6
C7H6F2O
144.121
MFCD01631340
YKCLAAQQWYPEDW-UHFFFAOYSA-N
2,6-difluoro-3-methyl-phenol,2,6-difluoro-m-cresol,acmc-209gp3,phenol, 2,6-difluoro-3-methyl,phenol, 2,6-difluoro-3-methyl-9ci
2774152
2,6-difluoro-3-methylphenol
CC1=C(C(=C(C=C1)F)O)F
261763-46-6
C7H6F2O
144.121
MFCD01631340
YKCLAAQQWYPEDW-UHFFFAOYSA-N
2,6-difluoro-3-methyl-phenol,2,6-difluoro-m-cresol,acmc-209gp3,phenol, 2,6-difluoro-3-methyl,phenol, 2,6-difluoro-3-methyl-9ci
2774152
2,6-difluoro-3-methylphenol
CC1=C(C(=C(C=C1)F)O)F
Quantity 1g
Boiling Point 182°C
UN Number UN3265
Formula Weight 144.12
Percent Purity ≥97%
Chemical Name or Material 2,6-Difluoro-3-methylphenol

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.