Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2,6-Difluoro-4-methoxybenzoic acid, 97%
Click to view available options
Quantity:
1g
10g
Specifications
Quantity | 1g |
Melting Point | 183°C to 185°C |
Formula Weight | 188.13 |
Percent Purity | 97% |
Chemical Name or Material | 2,6-Difluoro-4-methoxybenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures