Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',6'-Difluoroacetophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-7080621
Click to view available options
Quantity:
5 g
25 g
13670-99-0
C8H6F2O
156.13
MFCD00000328
VGIIILXIQLXVLC-UHFFFAOYSA-N
2',6'-difluoroacetophenone, 1-2,6-difluorophenyl ethanone, 2,6-difluoroacetophenone, 1-2,6-difluorophenyl ethan-1-one, ethanone, 1-2,6-difluorophenyl, acetophenone, 2',6'-difluoro, 1-acetyl-2,6-difluorobenzene, fr cf bv1, pubchem4218, intermediates-zcf02054
83643
CC(=O)C1=C(F)C=CC=C1F
13670-99-0
C8H6F2O
156.13
MFCD00000328
VGIIILXIQLXVLC-UHFFFAOYSA-N
2',6'-difluoroacetophenone, 1-2,6-difluorophenyl ethanone, 2,6-difluoroacetophenone, 1-2,6-difluorophenyl ethan-1-one, ethanone, 1-2,6-difluorophenyl, acetophenone, 2',6'-difluoro, 1-acetyl-2,6-difluorobenzene, fr cf bv1, pubchem4218, intermediates-zcf02054
83643
CC(=O)C1=C(F)C=CC=C1F

2?,6?-Difluoroacetophenone was used in the synthesis of 2-amino-4-alkyl- and 2-amino-4-arylquinazolines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,6′-Difluoroacetophenone was used in the synthesis of 2-amino-4-alkyl- and 2-amino-4-arylquinazolines.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.195
Boiling Point 76°C to 79°C (15 mmHg)
Flash Point 76°C (168°F)
Refractive Index 1.481
Quantity 5 g
Beilstein 2046068
Solubility Information Slightly soluble in water.
IUPAC Name 1-(2,6-difluorophenyl)ethanone
Formula Weight 156.13
Percent Purity 98%
Chemical Name or Material 2',6'-Difluoroacetophenone
Show More Show Less
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 237-151-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit