Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',6'-Difluoroacetophenone, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
13670-99-0
C8H6F2O
156.13
MFCD00000328
VGIIILXIQLXVLC-UHFFFAOYSA-N
2',6'-difluoroacetophenone, 1-2,6-difluorophenyl ethanone, 2,6-difluoroacetophenone, 1-2,6-difluorophenyl ethan-1-one, ethanone, 1-2,6-difluorophenyl, acetophenone, 2',6'-difluoro, 1-acetyl-2,6-difluorobenzene, fr cf bv1, pubchem4218, intermediates-zcf02054
83643
1-(2,6-difluorophenyl)ethanone
CC(=O)C1=C(F)C=CC=C1F
13670-99-0
C8H6F2O
156.13
MFCD00000328
VGIIILXIQLXVLC-UHFFFAOYSA-N
2',6'-difluoroacetophenone, 1-2,6-difluorophenyl ethanone, 2,6-difluoroacetophenone, 1-2,6-difluorophenyl ethan-1-one, ethanone, 1-2,6-difluorophenyl, acetophenone, 2',6'-difluoro, 1-acetyl-2,6-difluorobenzene, fr cf bv1, pubchem4218, intermediates-zcf02054
83643
1-(2,6-difluorophenyl)ethanone
CC(=O)C1=C(F)C=CC=C1F
Specific Gravity 1.19
Packaging Glass bottle
Refractive Index 1.48
Color Colorless to Yellow
Infrared Spectrum Authentic
Boiling Point 76.0°C to 79.0°C (15.0mmHg)
Quantity 5g
Density 1.1900g/mL
Flash Point 76°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula F2C6H3COCH3
Formula Weight 156.13
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2', 6'-Difluoroacetophenone
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 237-151-9