Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluoroanisole, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
437-82-1
C7H6F2O
144.12
IOBWAHRFIPQEQL-UHFFFAOYSA-N
2,6-difluoroanisole,1,3-difluoro-2-methoxy-benzene,benzene, 1,3-difluoro-2-methoxy,pubchem1968,2,6-difluoro anisole,2.6-difluoro anisole,acmc-1ajjj,ksc235o3h,iobwahrfipqeql-uhfffaoysa
2736897
1,3-difluoro-2-methoxybenzene
COC1=C(C=CC=C1F)F
437-82-1
C7H6F2O
144.12
IOBWAHRFIPQEQL-UHFFFAOYSA-N
2,6-difluoroanisole,1,3-difluoro-2-methoxy-benzene,benzene, 1,3-difluoro-2-methoxy,pubchem1968,2,6-difluoro anisole,2.6-difluoro anisole,acmc-1ajjj,ksc235o3h,iobwahrfipqeql-uhfffaoysa
2736897
1,3-difluoro-2-methoxybenzene
COC1=C(C=CC=C1F)F
Specific Gravity 1.221
Packaging Glass Bottle
Quantity 1g
Infrared Spectrum Authentic
Density 1.2210g/mL
Boiling Point 70.0°C to 72.0°C (56.0mmHg)
Flash Point 42°C
Assay Percent Range 97.5% min. (GC)
Formula Weight 144.12
Percent Purity 98%
Chemical Name or Material 2,6-Difluoroanisole
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Warning