Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Difluorobenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
162101-25-9
C6H5BF2O2
157.911
MFCD00792436
DBZAICSEFBVFHL-UHFFFAOYSA-N
2,6-difluorobenzeneboronic acid,2,6-difluorophenyl boronic acid,2,6-difluorophenyl boranediol,boronic acid, 2,6-difluorophenyl,2,6-difluorphenylboronic acid,2,6-difluoro phenylboric acid,boronic acid, b-2,6-difluorophenyl,2,6-difluorophenylboricacid,pubchem1816,acmc-1celt
2734336
(2,6-difluorophenyl)boronic acid
B(C1=C(C=CC=C1F)F)(O)O
162101-25-9
C6H5BF2O2
157.911
MFCD00792436
DBZAICSEFBVFHL-UHFFFAOYSA-N
2,6-difluorobenzeneboronic acid,2,6-difluorophenyl boronic acid,2,6-difluorophenyl boranediol,boronic acid, 2,6-difluorophenyl,2,6-difluorphenylboronic acid,2,6-difluoro phenylboric acid,boronic acid, b-2,6-difluorophenyl,2,6-difluorophenylboricacid,pubchem1816,acmc-1celt
2734336
(2,6-difluorophenyl)boronic acid
B(C1=C(C=CC=C1F)F)(O)O
Applications
2,6-Difluorobenzeneboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Quantity 25g
Melting Point ~140°C (decomposition)
Beilstein 8545931
Formula Weight 157.91
Percent Purity 98%
Chemical Name or Material 2,6-Difluorobenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning