Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluorobenzophenone, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
59189-51-4
C13H8F2O
218.203
MFCD00009895
KTRGJHDNHIPMEY-UHFFFAOYSA-N
2,6-difluorobenzophenone,2,6-difluorophenyl phenyl methanone,methanone, 2,6-difluorophenyl phenyl,2,6-difluorophenyl-phenylmethanone,2,6-difluorophenyl phenyl ketone,fr cf bvr,acmc-1ay87,2,6-difluorophenyl phenyl-methanone,2,6-difluorophenyl phenyl methanone #
100979
(2,6-difluorophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=C(C=CC=C2F)F
59189-51-4
C13H8F2O
218.203
MFCD00009895
KTRGJHDNHIPMEY-UHFFFAOYSA-N
2,6-difluorobenzophenone,2,6-difluorophenyl phenyl methanone,methanone, 2,6-difluorophenyl phenyl,2,6-difluorophenyl-phenylmethanone,2,6-difluorophenyl phenyl ketone,fr cf bvr,acmc-1ay87,2,6-difluorophenyl phenyl-methanone,2,6-difluorophenyl phenyl methanone #
100979
(2,6-difluorophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=C(C=CC=C2F)F
Density 1.2440g/mL
Quantity 5g
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula F2C6H3COC6H5
Formula Weight 218.2
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 2, 6-Difluorobenzophenone, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.