Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2,6-Difluorobenzophenone, 98%
Click to view available options
Quantity:
5g
25g
Specifications
Refractive Index | 1.5645 |
Density | 1.244 |
Quantity | 25g |
Boiling Point | 119°C to 120°C (1.5mmHg) |
Flash Point | >110°C (230°F) |
Beilstein | 2266589 |
Formula Weight | 218.2 |
Percent Purity | 98% |
Chemical Name or Material | 2,6-Difluorobenzophenone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 261-650-0
TSCA : No
Recommended Storage : Ambient temperatures