Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluorobenzoyl isocyanate, 80%, Tech., ACROS Organics™

Click to view available options
Quantity:
500mg
Packaging:
Glass bottle
60731-73-9
C8H3F2NO2
183.114
MFCD00673065
ZRJSABISRHPRSB-UHFFFAOYSA-N
2,6-difluorobenzoylisocyanate,pubchem16484,2.6-difluorobenzoyl isocyanate,2,6-difluoro-benzoyl isocyanate,benzoyl isocyanate, 2,6-difluoro,2,6-difluorophenyl oxomethanisocyanate,2,6-difluoro-n-oxomethylidene benzamide
2733369
2,6-difluorobenzoyl isocyanate
C1=CC(=C(C(=C1)F)C(=O)N=C=O)F
60731-73-9
C8H3F2NO2
183.114
MFCD00673065
ZRJSABISRHPRSB-UHFFFAOYSA-N
2,6-difluorobenzoylisocyanate,pubchem16484,2.6-difluorobenzoyl isocyanate,2,6-difluoro-benzoyl isocyanate,benzoyl isocyanate, 2,6-difluoro,2,6-difluorophenyl oxomethanisocyanate,2,6-difluoro-n-oxomethylidene benzamide
2733369
2,6-difluorobenzoyl isocyanate
C1=CC(=C(C(=C1)F)C(=O)N=C=O)F
Color White
Infrared Spectrum Authentic
Quantity 500mg
Packaging Glass bottle
CAS Min % 75.0
CAS Max % 100.0
Assay Percent Range 80%
Formula Weight 183.11
Physical Form Crystalline Solid
Percent Purity ≥75%
Grade Technical
Chemical Name or Material 2, 6-Difluorobenzoyl isocyanate, tech., 80%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning