Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Difluorobenzyl chloride, 98%

Click to view available options
Quantity:
1g
5g
697-73-4
C7H5ClF2
162.564
MFCD00191346
MJXRENZUAQXZGJ-UHFFFAOYSA-N
2,6-difluorobenzyl chloride,2-chloromethyl-1,3-difluorobenzene,2,6-difluorobenzylchloride,benzene, 2-chloromethyl-1,3-difluoro,alpha-chloro-2,6-difluorotoluene,2, 6-difluorobenzyl chloride,1-chloromethyl-2,6-difluorobenzene,2-chlormethyl-1,3-difluorbenzol,pubchem2178
581570
2-(chloromethyl)-1,3-difluorobenzene
C1=CC(=C(C(=C1)F)CCl)F
697-73-4
C7H5ClF2
162.564
MFCD00191346
MJXRENZUAQXZGJ-UHFFFAOYSA-N
2,6-difluorobenzyl chloride,2-chloromethyl-1,3-difluorobenzene,2,6-difluorobenzylchloride,benzene, 2-chloromethyl-1,3-difluoro,alpha-chloro-2,6-difluorotoluene,2, 6-difluorobenzyl chloride,1-chloromethyl-2,6-difluorobenzene,2-chlormethyl-1,3-difluorbenzol,pubchem2178
581570
2-(chloromethyl)-1,3-difluorobenzene
C1=CC(=C(C(=C1)F)CCl)F
Applications
2,6-Difluorobenzyl chloride is used as rufinamide intermediate. It is an important raw material and intermediate used in organic synthesis.

Solubility
Soluble in acetone, DMSO and DMF. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Quantity 5g
Boiling Point 172°C to 173°C (640mmHg)
Melting Point 32°C to 35°C
UN Number UN1760
Beilstein 243793
Solubility Information Soluble in acetone,DMSO and DMF. Insoluble in water.
Formula Weight 162.57
Percent Purity 98%
Chemical Name or Material 2,6-Difluorobenzyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H332-H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures