Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Difluorophenylacetonitrile, 98%

Click to view available options
Quantity:
1g
5g
654-01-3
C8H5F2N
153.132
MFCD00009973
GVAYBGQTAADLJS-UHFFFAOYSA-N
2,6-difluorophenylacetonitrile,2-2,6-difluorophenyl acetonitrile,2,6-difluorobenzyl cyanide,2,6-difluorophenyl acetonitrile,2,6-difluorobenzeneacetonitrile,benzeneacetonitrile, 2,6-difluoro,2-2,6-difluorophenyl ethanenitrile,pubchem4152,acmc-1axnw,n-dodecyl-beta-maltoside
69559
2-(2,6-difluorophenyl)acetonitrile
C1=CC(=C(C(=C1)F)CC#N)F
654-01-3
C8H5F2N
153.132
MFCD00009973
GVAYBGQTAADLJS-UHFFFAOYSA-N
2,6-difluorophenylacetonitrile,2-2,6-difluorophenyl acetonitrile,2,6-difluorobenzyl cyanide,2,6-difluorophenyl acetonitrile,2,6-difluorobenzeneacetonitrile,benzeneacetonitrile, 2,6-difluoro,2-2,6-difluorophenyl ethanenitrile,pubchem4152,acmc-1axnw,n-dodecyl-beta-maltoside
69559
2-(2,6-difluorophenyl)acetonitrile
C1=CC(=C(C(=C1)F)CC#N)F
Refractive Index 1.481
Density 1.238
Flash Point >100°C (212°F)
Quantity 5g
Boiling Point 109°C (27mmHg)
UN Number UN3276
Beilstein 2614807
Formula Weight 153.13
Percent Purity 98%
Chemical Name or Material 2,6-Difluorophenylacetonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c

H226-H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

EINECSNumber : 211-504-7

TSCA : No

Recommended Storage : Ambient temperatures