Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluorophenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
162101-25-9
C6H5BF2O2
157.911
DBZAICSEFBVFHL-UHFFFAOYSA-N
2,6-difluorobenzeneboronic acid,2,6-difluorophenyl boronic acid,2,6-difluorophenyl boranediol,boronic acid, 2,6-difluorophenyl,2,6-difluorphenylboronic acid,2,6-difluoro phenylboric acid,boronic acid, b-2,6-difluorophenyl,2,6-difluorophenylboricacid,pubchem1816,acmc-1celt
2734336
(2,6-difluorophenyl)boronic acid
B(C1=C(C=CC=C1F)F)(O)O
162101-25-9
C6H5BF2O2
157.911
DBZAICSEFBVFHL-UHFFFAOYSA-N
2,6-difluorobenzeneboronic acid,2,6-difluorophenyl boronic acid,2,6-difluorophenyl boranediol,boronic acid, 2,6-difluorophenyl,2,6-difluorphenylboronic acid,2,6-difluoro phenylboric acid,boronic acid, b-2,6-difluorophenyl,2,6-difluorophenylboricacid,pubchem1816,acmc-1celt
2734336
(2,6-difluorophenyl)boronic acid
B(C1=C(C=CC=C1F)F)(O)O
Color White
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 157.91
Physical Form Crystalline Powder
Percent Purity ≥96.5%
Chemical Name or Material 2, 6-Difluorophenylboronic acid
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.