Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Difluoropyridine-3-carboxaldehyde, 95%

Click to view available options
Quantity:
1g
5g
155601-65-3
C6H3F2NO
143.093
MFCD08277633
ZYCCUPSLEFLBNR-UHFFFAOYSA-N
2,6-difluoronicotinaldehyde,2,6-difluoropyridine-3-carboxaldehyde,3-pyridinecarboxaldehyde, 2,6-difluoro,2,6-difluoro-3-nicotinaldehyde,pubchem18894,2,6-difluoro-3-formylpyridine,2,6-difluoro-pyridine-3-carbaldehyde,2,6-difluoro-3-pyridinecarboxaldehyde,2,6-bis fluoranyl pyridine-3-carbaldehyde,3-pyridinecarboxaldehyde, 2,6-difluoro-9ci
10701929
2,6-difluoropyridine-3-carbaldehyde
C1=CC(=NC(=C1C=O)F)F
155601-65-3
C6H3F2NO
143.093
MFCD08277633
ZYCCUPSLEFLBNR-UHFFFAOYSA-N
2,6-difluoronicotinaldehyde,2,6-difluoropyridine-3-carboxaldehyde,3-pyridinecarboxaldehyde, 2,6-difluoro,2,6-difluoro-3-nicotinaldehyde,pubchem18894,2,6-difluoro-3-formylpyridine,2,6-difluoro-pyridine-3-carbaldehyde,2,6-difluoro-3-pyridinecarboxaldehyde,2,6-bis fluoranyl pyridine-3-carbaldehyde,3-pyridinecarboxaldehyde, 2,6-difluoro-9ci
10701929
2,6-difluoropyridine-3-carbaldehyde
C1=CC(=NC(=C1C=O)F)F
Sensitivity Moisture Sensitive
Refractive Index 1.4920
Quantity 5g
Boiling Point 64°C to 66°C (15mmHg)
Melting Point 19°C to 20°C
Density 1.378g/mL
Beilstein 7757317
Formula Weight 143.09
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 2,6-Difluoropyridine-3-carboxaldehyde
Show More Show Less

storageNote1 : -30° to -10°C