Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2,6-Difluoropyridine-3-carboxaldehyde, 95%
Click to view available options
Quantity:
1g
5g
Specifications
Refractive Index | 1.492 |
Sensitivity | Air sensitive |
Density | 1.378 g/mL |
Melting Point | 19°C to 20°C |
Quantity | 1g |
Boiling Point | 64°C to 66°C (15mmHg) |
Beilstein | 7757317 |
Formula Weight | 143.09 |
Physical Form | Liquid |
Percent Purity | 95% |
Show More |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C; Store under Argon