Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Difluoropyridine, 98+%

Click to view available options
Quantity:
5g
25g
100g
1513-65-1
C5H3F2N
115.083
MFCD00006227
MBTGBRYMJKYYOE-UHFFFAOYSA-N
pyridine, 2,6-difluoro,2,6-difluorpyridin,ccris 1720,2,6-difluoro-pyridine,2,6-difluoropyridin,pubchem9241,2,6 difluoropyridine,2, 6-difluoropyridine,2,6 difluoro pyridine,2,6-di-fluoropyridine
73934
2,6-difluoropyridine
C1=CC(=NC(=C1)F)F
1513-65-1
C5H3F2N
115.083
MFCD00006227
MBTGBRYMJKYYOE-UHFFFAOYSA-N
pyridine, 2,6-difluoro,2,6-difluorpyridin,ccris 1720,2,6-difluoro-pyridine,2,6-difluoropyridin,pubchem9241,2,6 difluoropyridine,2, 6-difluoropyridine,2,6 difluoro pyridine,2,6-di-fluoropyridine
73934
2,6-difluoropyridine
C1=CC(=NC(=C1)F)F
Applications
2,6-Difluoropyridine has been used in the preparation of poly(pyridine ether)s via polycondensation with silylated 1,1,1-tris(4-hydroxyphenyl)ethane.

Solubility
Not miscible or difficult to mix.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
Refractive Index 1.437
Density 1.268
Odor Characteristic
Boiling Point 123°C to 124°C
Flash Point 33°C (91°F)
Quantity 5g
UN Number UN1993
Beilstein 1422549
Solubility Information Not miscible or difficult to mix.
Formula Weight 115.08
Percent Purity ≥98%
Chemical Name or Material 2,6-Difluoropyridine
Show More Show Less

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 216-152-8

TSCA : No

Recommended Storage : Ambient temperatures