Alfa Aesar™ 2,6-Dihydroxy-4-methylpyridine, 98+%

Manufacturer: Alfa Aesar™ H27860MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27860MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4664-16-8
    Molecular Formula C6H7NO2
    Formula Weight 125.13
    MDL Number MFCD00040464
    Synonym 2,6-Dihydroxy-4-picoline; 6-Hydroxy-4-methyl-2-pyridone
    Chemical Name or Material 2,6-Dihydroxy-4-methylpyridine

    Specifications

    Formula Weight 125.13
    Molecular Formula C6H7NO2
    MDL Number MFCD00040464
    CAS 4664-16-8
    Synonym 2,6-Dihydroxy-4-picoline; 6-Hydroxy-4-methyl-2-pyridone
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 195° to 198°C
    Quantity 250mg
    Chemical Name or Material 2,6-Dihydroxy-4-methylpyridine
    Assay ≥98%

    Description

    Applications
    In order to synthesize macrocycles containing pyridine, 2,6-his-(bromomethyl)pyridine was prepared by bromonating 2,6-di (hydroxymethyl)pyridine.

    Solubility
    Soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit