Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dihydroxyacetophenone, Matrix Substance for MALDI-MS, ≥99.5%, MilliporeSigma™ Supelco™

$72.90 - $288.00

Specifications

Percent Purity ≥99.5% (HPLC); ≥99.5%
Grade Matrix Substance for MALDI-MS
Linear Formula (HO)2C6H3 COCH3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-5280
View Documents
MilliporeSigma Supelco
374681GF
1 g
Each for $72.90
Add to cart
 
11-101-5281
View Documents
MilliporeSigma Supelco
374685GF
5 g
Each for $288.00
Add to cart
 
Description

Description

Specifications

Specifications

≥99.5% (HPLC); ≥99.5%
Matrix Substance for MALDI-MS
(HO)2C6H3 COCH3
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit