Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2,6-Diisopropylphenyl isocyanate, 94%
Click to view available options
Quantity:
1g
5g
25g
Specifications
Quantity | 1g |
Refractive Index | 1.518 |
Density | 0.951 |
Sensitivity | Moisture sensitive |
Boiling Point | 116°C to 117°C (10mmHg) |
Flash Point | 107°C (224°F) |
UN Number | UN2206 |
Beilstein | 2645721 |
Formula Weight | 203.29 |
Percent Purity | 94% |
Show More |
Safety and Handling
GHS H Statement
H330-H334-H318-H302-H315-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H330-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber : 248-885-4
RTECSNumber : CY8545000
TSCA : Yes
Recommended Storage : Keep cold