Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Dimethoxybenzeneboronic acid, 97+%

Click to view available options
Quantity:
1g
5g
25g
23112-96-1
C8H11BO4
181.982
MFCD01318987
BKWVXPCYDRURMK-UHFFFAOYSA-N
2,6-dimethoxyphenyl boronic acid,2,6-dimethoxyphenylboronicacid,2,6-dimethoxybenzeneboronic acid,2,6-dimethoxyphenyl-boronic acid,2,6-dimethoxyphenyl boranediol,boronic acid, 2,6-dimethoxyphenyl,pubchem1825,acmc-1clhv,ksc201q0f,2,6-dimethoxy-phenylboronic acid
2734343
(2,6-dimethoxyphenyl)boronic acid
B(C1=C(C=CC=C1OC)OC)(O)O
23112-96-1
C8H11BO4
181.982
MFCD01318987
BKWVXPCYDRURMK-UHFFFAOYSA-N
2,6-dimethoxyphenyl boronic acid,2,6-dimethoxyphenylboronicacid,2,6-dimethoxybenzeneboronic acid,2,6-dimethoxyphenyl-boronic acid,2,6-dimethoxyphenyl boranediol,boronic acid, 2,6-dimethoxyphenyl,pubchem1825,acmc-1clhv,ksc201q0f,2,6-dimethoxy-phenylboronic acid
2734343
(2,6-dimethoxyphenyl)boronic acid
B(C1=C(C=CC=C1OC)OC)(O)O
Quantity 1g
Melting Point 106°C to 108°C
Beilstein 3032352
Formula Weight 181.98
Percent Purity ≥97%
Chemical Name or Material 2,6-Dimethoxybenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning