Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxybenzoic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
1466-76-8
C9H10O4
182.18
MFCD00002437
MBIZFBDREVRUHY-UHFFFAOYSA-N
benzoic acid, 2,6-dimethoxy,2,6-dimethoxy-benzoic acid,unii-g6b3p1lo43,2,6-dimethoxybenzoicacid,pubchem19743,acmc-1bwjj,intermediates-zcf02125,benzoic acid,6-dimethoxy,dsstox_cid_26999,dsstox_rid_82043
15109
2,6-dimethoxybenzoic acid
COC1=C(C(=CC=C1)OC)C(=O)O
1466-76-8
C9H10O4
182.18
MFCD00002437
MBIZFBDREVRUHY-UHFFFAOYSA-N
benzoic acid, 2,6-dimethoxy,2,6-dimethoxy-benzoic acid,unii-g6b3p1lo43,2,6-dimethoxybenzoicacid,pubchem19743,acmc-1bwjj,intermediates-zcf02125,benzoic acid,6-dimethoxy,dsstox_cid_26999,dsstox_rid_82043
15109
2,6-dimethoxybenzoic acid
COC1=C(C(=CC=C1)OC)C(=O)O
Melting Point 187°C to 190°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color White
Quantity 100g
Water 0.5% max. (K.F.)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3O)2C6H3CO2H
Beilstein 10, 388
Solubility Information Solubility in water: 0,35 wt% (25 c). Other solubilities: 0.29 wt% in benzene (25 c),3.64 wt% in chloroform (25 c)
Formula Weight 182.18
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 2, 6-Dimethoxybenzoic acid, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 215-985-4

RTECSNumber : DG8598725

TSCA : TSCA