Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxybenzonitrile, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
16932-49-3
C9H9NO2
163.18
MFCD00001788
XHAHKSSLDJIEDH-UHFFFAOYSA-N
benzonitrile, 2,6-dimethoxy,2,6-dimethoxy-benzonitrile,2,6-dimethoxybenzenecarbonitrile,pubchem4772,2.6-dimethoxybenzonitrile,benzonitrile,2,6-dimethoxy,2-10-00-00260 beilstein handbook reference,2,6-dimethoxybenzonitrile
85648
2,6-dimethoxybenzonitrile
COC1=C(C(=CC=C1)OC)C#N
16932-49-3
C9H9NO2
163.18
MFCD00001788
XHAHKSSLDJIEDH-UHFFFAOYSA-N
benzonitrile, 2,6-dimethoxy,2,6-dimethoxy-benzonitrile,2,6-dimethoxybenzenecarbonitrile,pubchem4772,2.6-dimethoxybenzonitrile,benzonitrile,2,6-dimethoxy,2-10-00-00260 beilstein handbook reference,2,6-dimethoxybenzonitrile
85648
2,6-dimethoxybenzonitrile
COC1=C(C(=CC=C1)OC)C#N
Boiling Point 310°C
Melting Point 119°C to 123°C
Infrared Spectrum Authentic
Color Beige
Quantity 10g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CH3O)2C6H3CN
Beilstein 10, 388
Formula Weight 163.18
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2, 6-Dimethoxybenzonitrile
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 241-000-2