Alfa Aesar™ 2,6-Dimethoxyphenol, 99%

Manufacturer: Alfa Aesar™ B2425314

Quantity 25g

 View more versions of this product

Catalog No. AAB2425314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 91-10-1
    Molecular Formula C8H10O3
    Formula Weight 154.16
    MDL Number MFCD00064434
    Beilstein 1526871
    Synonym Pyrogallol 1,3-dimethyl ether; Syringol
    Chemical Name or Material 2,6-Dimethoxyphenol

    Specifications

    Formula Weight 154.16
    TSCA TSCA
    Molecular Formula C8H10O3
    MDL Number MFCD00064434
    CAS 91-10-1
    Beilstein 1526871
    Synonym Pyrogallol 1,3-dimethyl ether; Syringol
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number SL0900000
    Flash Point 140°C (284°F)
    Boiling Point 261°C
    Melting Point 50° to 57°C
    Quantity 25g
    Chemical Name or Material 2,6-Dimethoxyphenol
    Assay 99%

    Description

    Applications
    It is used in smoke flavors, whisky, rum, tea, spice, savory, seafood, meat, liquorices, coffee, and nut flavors. It is reported to be the single most important flavor chemical in smoke flavors.

    Solubility
    Soluble in water.

    Notes
    Store at room temperature. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit