Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxyphenol, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
91-10-1
C8H10O3
154.17
MFCD00064434
KLIDCXVFHGNTTM-UHFFFAOYSA-N
syringol,pyrogallol 1,3-dimethyl ether,1,3-dimethyl pyrogallate,phenol, 2,6-dimethoxy,2-hydroxy-1,3-dimethoxybenzene,1,3-di-o-methylpyrogallol,1,3-dimethoxy-2-hydroxybenzene,2,6-dimethoxy-phenol,pyrogallol dimethylether,2,6-dwumetoksyfenol
7041
CHEBI:955
2,6-dimethoxyphenol
COC1=C(C(=CC=C1)OC)O
91-10-1
C8H10O3
154.17
MFCD00064434
KLIDCXVFHGNTTM-UHFFFAOYSA-N
syringol,pyrogallol 1,3-dimethyl ether,1,3-dimethyl pyrogallate,phenol, 2,6-dimethoxy,2-hydroxy-1,3-dimethoxybenzene,1,3-di-o-methylpyrogallol,1,3-dimethoxy-2-hydroxybenzene,2,6-dimethoxy-phenol,pyrogallol dimethylether,2,6-dwumetoksyfenol
7041
CHEBI:955
2,6-dimethoxyphenol
COC1=C(C(=CC=C1)OC)O
Melting Point 50°C to 56°C
Boiling Point 261°C
Flash Point 140°C
Quantity 25g
Color Brown to Gray or White
Infrared Spectrum Authentic
Packaging Glass bottle
Water 0.5% max.
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3O)2C6H3OH
Beilstein 06,1081
Solubility Information Solubility in water: 2% (13°C). Other solubilities: soluble in ethyl ether,ethanol
Formula Weight 154.17
Physical Form Crystalline Powder, Crystals, or Crystalline Solid
Percent Purity ≥98.5%
Chemical Name or Material 2, 6-Dimethoxyphenol, 99+%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-041-1

RTECSNumber : SL0900000

TSCA : TSCA