Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxyphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
23112-96-1
C8H11BO4
181.982
MFCD01318987
BKWVXPCYDRURMK-UHFFFAOYSA-N
2,6-dimethoxyphenyl boronic acid,2,6-dimethoxyphenylboronicacid,2,6-dimethoxybenzeneboronic acid,2,6-dimethoxyphenyl-boronic acid,2,6-dimethoxyphenyl boranediol,boronic acid, 2,6-dimethoxyphenyl,pubchem1825,acmc-1clhv,ksc201q0f,2,6-dimethoxy-phenylboronic acid
2734343
(2,6-dimethoxyphenyl)boronic acid
B(C1=C(C=CC=C1OC)OC)(O)O
23112-96-1
C8H11BO4
181.982
MFCD01318987
BKWVXPCYDRURMK-UHFFFAOYSA-N
2,6-dimethoxyphenyl boronic acid,2,6-dimethoxyphenylboronicacid,2,6-dimethoxybenzeneboronic acid,2,6-dimethoxyphenyl-boronic acid,2,6-dimethoxyphenyl boranediol,boronic acid, 2,6-dimethoxyphenyl,pubchem1825,acmc-1clhv,ksc201q0f,2,6-dimethoxy-phenylboronic acid
2734343
(2,6-dimethoxyphenyl)boronic acid
B(C1=C(C=CC=C1OC)OC)(O)O
Quantity 5g
Packaging Glass bottle
Color White to Beige
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 181.98
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 2, 6-Dimethoxyphenylboronic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.