Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxypyridine 99%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
6231-18-1
C7H9NO2
139.154
MFCD00006266
IBTGEEMBZJBBSH-UHFFFAOYSA-N
pyridine, 2,6-dimethoxy,2,6-dimethoxy pyridine,pyridine,2,6-dimethoxy,acmc-1b7tt,ksc494c3n,2,6-dimethoxypyridine,2,6-dimethoxypyridine 25g
80378
2,6-dimethoxypyridine
COC1=NC(=CC=C1)OC
6231-18-1
C7H9NO2
139.154
MFCD00006266
IBTGEEMBZJBBSH-UHFFFAOYSA-N
pyridine, 2,6-dimethoxy,2,6-dimethoxy pyridine,pyridine,2,6-dimethoxy,acmc-1b7tt,ksc494c3n,2,6-dimethoxypyridine,2,6-dimethoxypyridine 25g
80378
2,6-dimethoxypyridine
COC1=NC(=CC=C1)OC
Density 1.053g/mL
Packaging Glass bottle
Quantity 10g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (C5H3N)(OCH3)2
Formula Weight 139.15
Chemical Name or Material 2, 6-Dimethoxypyridine, 99%

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 228-334-4

TSCA : TSCA