Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
269410-25-5
C14H21BO3
248.13
MFCD02093724
TYCKOBOJYNRIBO-UHFFFAOYSA-N
2,6-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol,4-hydroxy-3,5-dimethylphenylboronic acid pinacol ester,3,5-dimethyl-4-hydroxyphenylboronic acid pinacol ester,3,5-dimethyl-4-hydroxybenzeneboronic acid, pinacol ester,4-hydroxy-3,5-dimethylphenylboronic acid, pinacol ester,2,6-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol,2,6-dimethyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenol,2,6-dimethyl-4-4,4-5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol,2-4-hydroxy-3,5-dimethylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,phenol, 2,6-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
2734649
2,6-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)C)O)C
269410-25-5
C14H21BO3
248.13
MFCD02093724
TYCKOBOJYNRIBO-UHFFFAOYSA-N
2,6-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol,4-hydroxy-3,5-dimethylphenylboronic acid pinacol ester,3,5-dimethyl-4-hydroxyphenylboronic acid pinacol ester,3,5-dimethyl-4-hydroxybenzeneboronic acid, pinacol ester,4-hydroxy-3,5-dimethylphenylboronic acid, pinacol ester,2,6-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol,2,6-dimethyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenol,2,6-dimethyl-4-4,4-5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol,2-4-hydroxy-3,5-dimethylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,phenol, 2,6-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
2734649
2,6-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)C)O)C
Infrared Spectrum Authentic
Quantity 1g
Color White to Beige
Packaging Glass bottle
Melting Point 104.0°C to 109.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: insoluble in water
Formula Weight 248.13
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2,6-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C