Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethyl-4-heptanone, Remainder Mainly4,6-dimethyl-2-heptanone (Tech.), 80%, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
20L
Packaging:
Glass Bottle
Metal drum
108-83-8,19549-80-5
C9H18O
142.242
MFCD00008940
PTTPXKJBFFKCEK-UHFFFAOYSA-N
2,6-dimethyl-4-heptanone,diisobutyl ketone,isovalerone,isobutyl ketone,valerone,diisobutylketone,diisobutilchetone,s-diisopropylacetone,di-isobutylcetone,dibk
7958
2,6-dimethylheptan-4-one
CC(C)CC(=O)CC(C)C
108-83-8,19549-80-5
C9H18O
142.242
MFCD00008940
PTTPXKJBFFKCEK-UHFFFAOYSA-N
2,6-dimethyl-4-heptanone,diisobutyl ketone,isovalerone,isobutyl ketone,valerone,diisobutylketone,diisobutilchetone,s-diisopropylacetone,di-isobutylcetone,dibk
7958
2,6-dimethylheptan-4-one
CC(C)CC(=O)CC(C)C
Infrared Spectrum Authentic
Water 0.15% max. (K.F.)
Color Colorless to Yellow
Packaging Glass Bottle
Quantity 2.5L
Refractive Index 1.4120 to 1.4150 (20°C, 589nm)
Density 0.8090g/mL
CAS Min % 75.0
CAS Max % 100.0
Assay Percent Range 80%
Linear Formula (CH3)2CHCH2COCH2CH(CH3)2
Beilstein 01, 710
Formula Weight 142.24
Physical Form Liquid
Percent Purity ≥75% (typically) exact percentage on the certificate of analysis, 96% min. (main compound plus isomer)
Grade Technical
Chemical Name or Material 2, 6-Dimethyl-4-heptanone, tech., 80%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 203-620-1

RTECSNumber : MJ5775000

TSCA : TSCA