Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethyl-γ-pyrone, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
1004-36-0
C7H8O2
124.14
MFCD00006579
VSYFZULSKMFUJJ-UHFFFAOYSA-N
2,6-dimethyl-4h-pyran-4-one,2,6-dimethyl-4-pyrone,2,6-dimethyl-gamma-pyrone,2,6-dimethyl-4-pyranone,4h-pyran-4-one, 2,6-dimethyl,gamma-2,6-dimethylpyrone,unii-v2d262mf0l,2,6-dimethyl-pyran-4-one,2,6-dimethyl-.gamma.-pyrone,ccris 3601
13862
2,6-dimethylpyran-4-one
CC1=CC(=O)C=C(O1)C
1004-36-0
C7H8O2
124.14
MFCD00006579
VSYFZULSKMFUJJ-UHFFFAOYSA-N
2,6-dimethyl-4h-pyran-4-one,2,6-dimethyl-4-pyrone,2,6-dimethyl-gamma-pyrone,2,6-dimethyl-4-pyranone,4h-pyran-4-one, 2,6-dimethyl,gamma-2,6-dimethylpyrone,unii-v2d262mf0l,2,6-dimethyl-pyran-4-one,2,6-dimethyl-.gamma.-pyrone,ccris 3601
13862
2,6-dimethylpyran-4-one
CC1=CC(=O)C=C(O1)C
Boiling Point 248°C to 250°C
Quantity 500g
Melting Point 133°C to 137°C
Infrared Spectrum Authentic
Color White to Beige
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 17, 291
Formula Weight 124.14
Physical Form Crystalline Powder or Crystals
Percent Purity ≥98.5%
Chemical Name or Material 2,6-Dimethyl-γ-pyrone
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.