Learn More
eMolecules​ 2,6-Dimethylacetophenone | 2142-76-9 | 1G | Purity: 97% DFS Item
Manufacturer:  eMolecules​ 249361451
Combi-Blocks | 2,6-Dimethylacetophenone | 1G | 2142-76-9 | MFCD01861394
Catalog No. 50-158-0188
Due to product restrictions, please to purchase or view availability for this product.