Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylaniline 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
87-62-7
C8H11N
121.18
MFCD00007747
UFFBMTHBGFGIHF-UHFFFAOYSA-N
2,6-xylidine,2-amino-m-xylene,o-xylidine,2,6-dimethylbenzenamine,2-amino-1,3-dimethylbenzene,2,6-dimethylphenylamine,benzenamine, 2,6-dimethyl,2,6-xylylamine,2-amino-1,3-xylene,1-amino-2,6-dimethylbenzene
6896
CHEBI:28738
2,6-dimethylaniline
CC1=C(C(=CC=C1)C)N
87-62-7
C8H11N
121.18
MFCD00007747
UFFBMTHBGFGIHF-UHFFFAOYSA-N
2,6-xylidine,2-amino-m-xylene,o-xylidine,2,6-dimethylbenzenamine,2-amino-1,3-dimethylbenzene,2,6-dimethylphenylamine,benzenamine, 2,6-dimethyl,2,6-xylylamine,2-amino-1,3-xylene,1-amino-2,6-dimethylbenzene
6896
CHEBI:28738
2,6-dimethylaniline
CC1=C(C(=CC=C1)C)N
Boiling Point 216°C
Melting Point 10°C to 12°C
Flash Point 91°C
Specific Gravity 0.98
Refractive Index 1.5591 to 1.5611
Quantity 5g
Color Undesignated
Packaging Glass bottle
Density 0.9800g/mL
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2C6H3NH2
Beilstein 12, 1107
Merck Index 15, 10282
Solubility Information Solubility in water: 7.5g/L (20 c). Other solubilities: very soluble in ether and ethanol,soluble in dimethyl sulfoxide,carbon tetrachlorid
Formula Weight 121.18
Physical Form Liquid
Percent Purity 98.5 to 101.5% total base, ≥98.5% (GC)
Chemical Name or Material 2, 6-Dimethylaniline, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-758-7

RTECSNumber : ZE9275000

TSCA : TSCA

Recommended Storage : product darkens if exposed to light and air