Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylbenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number AAH6450322
Click to view available options
Quantity:
5 g
25 g
100 g
1123-56-4
C9H10O
134.178
MFCD00128003
QOJQBWSZHCKOLL-UHFFFAOYSA-N
m-xylene-2-carboxaldehyde, 2,6-dimethyl-benzaldehyde, 2-formyl-m-xylene, benzaldehyde, 2,6-dimethyl, 2,6-dimethylbenzaldehyd, acmc-1bown, 2,6-dimethyl benzaldehyde, ksc493s1p, pharmabridge p-2713, 2,6-dimethylbenzaldehyde
583841
2,6-dimethylbenzaldehyde
CC1=C(C(=CC=C1)C)C=O
1123-56-4
C9H10O
134.178
MFCD00128003
QOJQBWSZHCKOLL-UHFFFAOYSA-N
m-xylene-2-carboxaldehyde, 2,6-dimethyl-benzaldehyde, 2-formyl-m-xylene, benzaldehyde, 2,6-dimethyl, 2,6-dimethylbenzaldehyd, acmc-1bown, 2,6-dimethyl benzaldehyde, ksc493s1p, pharmabridge p-2713, 2,6-dimethylbenzaldehyde
583841
2,6-dimethylbenzaldehyde
CC1=C(C(=CC=C1)C)C=O

An aldehyde containing building block used for proteomics research

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An aldehyde containing building block used for proteomics research

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 27°C to 30°C (literature)
Color White to Yellow
Boiling Point 230°C (literature)
Flash Point 96°C (205°F)
Refractive Index 1.548
Quantity 100 g
Beilstein 907019
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 134.18
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,6-Dimethylbenzaldehyde
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit