Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Dimethylbenzeneboronic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
100379-00-8
C8H11BO2
149.984
MFCD01009693
ZXDTWWZIHJEZOG-UHFFFAOYSA-N
2,6-dimethylphenyl boronic acid,2,6-dimethylbenzeneboronic acid,2,6-dimethylphenyl boranediol,2,6-dimethylboronic acid,m-xylene-2-boronic acid,2,6-dimethyl-phenylboronic acid,boronic acid, 2,6-dimethylphenyl,2-borono-m-xylene,pubchem16357,acmc-1c3za
583322
(2,6-dimethylphenyl)boronic acid
B(C1=C(C=CC=C1C)C)(O)O
100379-00-8
C8H11BO2
149.984
MFCD01009693
ZXDTWWZIHJEZOG-UHFFFAOYSA-N
2,6-dimethylphenyl boronic acid,2,6-dimethylbenzeneboronic acid,2,6-dimethylphenyl boranediol,2,6-dimethylboronic acid,m-xylene-2-boronic acid,2,6-dimethyl-phenylboronic acid,boronic acid, 2,6-dimethylphenyl,2-borono-m-xylene,pubchem16357,acmc-1c3za
583322
(2,6-dimethylphenyl)boronic acid
B(C1=C(C=CC=C1C)C)(O)O
Applications
It is used as a reagent for palladium catalyzed Suzuki-Miyaura coupling reactions, Pd-catalyzed homocouplings, Rhodium(I)-catalyzed 1,4-addition reactions. It is used in the preparation of protein kinase inhibitors, G Protein-Coupled Receptor.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Quantity 5g
Melting Point 120°C to 126°C (decomposition)
Formula Weight 149.99
Percent Purity 97%
Chemical Name or Material 2,6-Dimethylbenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning