Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylbenzyl chloride, 98%, Thermo Scientific™

Click to view available options
Quantity:
5g
25g
100g
5402-60-8
C9H11Cl
154.64
HPVRFWQMBYLJRL-UHFFFAOYSA-N
2,6-dimethylbenzyl chloride, 2-chloromethyl-1,3-dimethylbenzene, 2,6-dimethylbenzylchloride, 2,6-dimethyl benzylchloride, 1-chloromethyl-2,6-dimethylbenzene, 2-chloromethyl-1,3-dimethyl-benzene, 2-chloromethyl-m-xylene, benzene, 2-chloromethyl-1,3-dimethyl, 2, 6-dimethylbenzyl chloride
221154
2-(chloromethyl)-1,3-dimethylbenzene
CC1=C(C(=CC=C1)C)CCl
5402-60-8
C9H11Cl
154.64
HPVRFWQMBYLJRL-UHFFFAOYSA-N
2,6-dimethylbenzyl chloride, 2-chloromethyl-1,3-dimethylbenzene, 2,6-dimethylbenzylchloride, 2,6-dimethyl benzylchloride, 1-chloromethyl-2,6-dimethylbenzene, 2-chloromethyl-1,3-dimethyl-benzene, 2-chloromethyl-m-xylene, benzene, 2-chloromethyl-1,3-dimethyl, 2, 6-dimethylbenzyl chloride
221154
2-(chloromethyl)-1,3-dimethylbenzene
CC1=C(C(=CC=C1)C)CCl
Boiling Point 100°C (2.0 kPa)
Melting Point 33.0°C
Quantity 100g
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Formula Weight 154.64
Percent Purity 98%
Chemical Name or Material 2,6-Dimethylbenzyl chloride

GHS H Statement May be corrosive to metals. Causes severe skin burns and eye damage.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

RUO – Research Use Only