Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylcyclohexanol, Mixture of Isomers 99%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
Packaging:
Glass bottle
5337-72-4
C8H16O
128.21
MFCD00001502
MOISVRZIQDQVPF-UHFFFAOYSA-N
2,6-dimethylcyclohexanol,cyclohexanol, 2,6-dimethyl,2,6-dimethylcyclohexyl alcohol,ambsclk-255,cyclohexanol,6-dimethyl,2,6-dimethyl-cyclohexanol,acmc-20ao81,2,6-dimethylcyclohexanol,c&t,2,6-dimethyl-cyclohexan-1-ol,2,6-dimethylcyclohexanol, mixture of isomers
21428
2,6-dimethylcyclohexan-1-ol
CC1CCCC(C1O)C
5337-72-4
C8H16O
128.21
MFCD00001502
MOISVRZIQDQVPF-UHFFFAOYSA-N
2,6-dimethylcyclohexanol,cyclohexanol, 2,6-dimethyl,2,6-dimethylcyclohexyl alcohol,ambsclk-255,cyclohexanol,6-dimethyl,2,6-dimethyl-cyclohexanol,acmc-20ao81,2,6-dimethylcyclohexanol,c&t,2,6-dimethyl-cyclohexan-1-ol,2,6-dimethylcyclohexanol, mixture of isomers
21428
2,6-dimethylcyclohexan-1-ol
CC1CCCC(C1O)C
Refractive Index 1.4585 to 1.4615
Density 0.9400g/mL
Flash Point 55°C
Boiling Point 174.4°C
Specific Gravity 0.94
Quantity 5g
Color Colorless
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2C6H9OH
Beilstein 06, 18
Formula Weight 128.21
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2, 6-Dimethylcyclohexanol, 99%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-264-9

TSCA : TSCA