Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylcyclohexanone, cis + trans, 99%, Thermo Scientific™

Click to view available options
Quantity:
25g
100g
500g
2816-57-1
C8H14O
126.199
MFCD00001637
AILVYPLQKCQNJC-UHFFFAOYSA-N
2,6-dimethylcyclohexanone, cyclohexanone, 2,6-dimethyl, cyclohexanone,6-dimethyl, 2,6-dimethyl cyclohexanone, 2,6-dimethyl-cyclohexanone, cyclohexanone,2,6-dimethyl, 2,6-dimethylcyclohexanone,c&t, 2,6-dimethyl-cyclohexan-1-one, 2,6-dimethylcyclohexanone cis + trans, 2,6-dimethylcyclohexanone, cis + trans
17780
2,6-dimethylcyclohexan-1-one
CC1CCCC(C1=O)C
2816-57-1
C8H14O
126.199
MFCD00001637
AILVYPLQKCQNJC-UHFFFAOYSA-N
2,6-dimethylcyclohexanone, cyclohexanone, 2,6-dimethyl, cyclohexanone,6-dimethyl, 2,6-dimethyl cyclohexanone, 2,6-dimethyl-cyclohexanone, cyclohexanone,2,6-dimethyl, 2,6-dimethylcyclohexanone,c&t, 2,6-dimethyl-cyclohexan-1-one, 2,6-dimethylcyclohexanone cis + trans, 2,6-dimethylcyclohexanone, cis + trans
17780
2,6-dimethylcyclohexan-1-one
CC1CCCC(C1=O)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Density 0.918
Boiling Point 174°C to 176°C
Flash Point 51°C (123°F)
UN Number UN1224
Beilstein 1099000
Refractive Index 1.447
Quantity 25g
Formula Weight 126.2
Percent Purity 99%
Chemical Name or Material 2,6-Dimethylcyclohexanone, cis + trans

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 220-570-6

TSCA : No

Recommended Storage : Ambient temperatures

RUO – Research Use Only