Learn More
eMolecules​ 2,6-Dimethylisonicotinic acid | 54221-93-1 | 1G | Purity: 98% DFS Item
Manufacturer:  eMolecules​ 205409298
Combi-Blocks | 2,6-Dimethylisonicotinic acid | 1G | 54221-93-1 | MFCD03731176
Catalog No. 50-157-8055
Due to product restrictions, please to purchase or view availability for this product.