Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Dimethylmorpholine, cis + trans, 97%

Manufacturer:  Alfa Aesar™ H2595414

Quantity 25g

 View more versions of this product

Catalog No. AAH2595414


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

141-91-3
115.18
UN1992
2,6-Dimethylmorpholine, cis + trans
C6H13NO
MFCD00005973
103036

Specifications

141-91-3
−85°C
115.18
UN1992
25g
2,6-Dimethylmorpholine, cis + trans
C6H13NO
147°C
MFCD00005973
103036
48°C (120°F)
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318-H226-H312
Causes serious eye damage.
Flammable liquid and vapour.
Harmful in contact with skin.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

RTECSNumber : QE1750000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit