Alfa Aesar™ 2,6-Dimethylmorpholine, cis + trans, 97%

Manufacturer: Alfa Aesar™ H2595422

Quantity 100g

 View more versions of this product

Catalog No. AAH2595422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 141-91-3
    Molecular Formula C6H13NO
    Formula Weight 115.18
    MDL Number MFCD00005973
    UN Number UN1992
    Beilstein 103036
    Chemical Name or Material 2,6-Dimethylmorpholine, cis + trans

    Specifications

    Formula Weight 115.18
    Melting Point −85°C
    MDL Number MFCD00005973
    Flash Point 48°C (120°F)
    Health Hazard 1 Danger
    Molecular Formula C6H13NO
    Boiling Point 147°C
    UN Number UN1992
    Quantity 100g
    Health Hazard 2 GHS H Statement
    H318-H226-H312
    Causes serious eye damage.
    Flammable liquid and vapour.
    Harmful in contact with skin.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
    RTECS Number QE1750000
    TSCA TSCA
    CAS 141-91-3
    Beilstein 103036
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2,6-Dimethylmorpholine, cis + trans
    Assay 97%

    GHS H Statement
    H318-H226-H312
    Causes serious eye damage.
    Flammable liquid and vapour.
    Harmful in contact with skin.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit