Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylphenylmagnesium bromide, 1M solution in THF, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
21450-64-6,109-99-9
C8H9BrMg
209.369
DCQJQVFIEFMTTQ-UHFFFAOYSA-M
2,6-dimethylphenylmagnesium bromide,2,6-dimethylphenylmagnesium bromide solution,biguvduzgajwqk-uhfffaoysa-m,2,6-dimethylphenymagnesium bromide,bromo 2,6-dimethylphenyl magnesium,2,6-dimethyl-phenylmagnesium bromide,2,6-dimethylphenyl magnesium bromide,2,6-dimethylphenylmagnesium bromide, 0.5m in 2-methf,2,6-dimethylphenylmagnesium bromide 0.5 m in tetrahydrofuran,2,6-dimethylphenylmagnesium bromide solution, 1.0 m in thf
4517616
magnesium;1,3-dimethylbenzene-2-ide;bromide
CC1=[C-]C(=CC=C1)C.[Mg+2].[Br-]
21450-64-6,109-99-9
C8H9BrMg
209.369
DCQJQVFIEFMTTQ-UHFFFAOYSA-M
2,6-dimethylphenylmagnesium bromide,2,6-dimethylphenylmagnesium bromide solution,biguvduzgajwqk-uhfffaoysa-m,2,6-dimethylphenymagnesium bromide,bromo 2,6-dimethylphenyl magnesium,2,6-dimethyl-phenylmagnesium bromide,2,6-dimethylphenyl magnesium bromide,2,6-dimethylphenylmagnesium bromide, 0.5m in 2-methf,2,6-dimethylphenylmagnesium bromide 0.5 m in tetrahydrofuran,2,6-dimethylphenylmagnesium bromide solution, 1.0 m in thf
4517616
magnesium;1,3-dimethylbenzene-2-ide;bromide
CC1=[C-]C(=CC=C1)C.[Mg+2].[Br-]
CAS Min % 19.93,76.92
CAS Max % 23.08,80.07
Solubility Information Solubility in water: reacts.
Formula Weight 209.37
Physical Form Solution
Concentration 0.95 to 1.10M
Density 0.9980g/mL
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: reacts.
Specific Gravity 0.998
Boiling Point 65.0°C to 67.0°C
Color Brown-Yellow to Colorless
Quantity 100mL
Flash Point −19°C
Chemical Name or Material 2,6-Dimethylphenylmagnesium bromide
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor. Causes severe skin burns and eye damage. May cause respiratory irritation. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger