Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Dimethylpiperidine, predominantly cis, 99%

Click to view available options
Quantity:
5g
50g
504-03-0
C7H15N
113.204
MFCD00066304
SDGKUVSVPIIUCF-UHFFFAOYSA-N
nanofin,lupetidine,naniopinum,2,6-lupetidine,lupetidin,nanophin,nanophine,nanophyn,piperidine, 2,6-dimethyl,naniopine
68843
2,6-dimethylpiperidine
CC1CCCC(N1)C
504-03-0
C7H15N
113.204
MFCD00066304
SDGKUVSVPIIUCF-UHFFFAOYSA-N
nanofin,lupetidine,naniopinum,2,6-lupetidine,lupetidin,nanophin,nanophine,nanophyn,piperidine, 2,6-dimethyl,naniopine
68843
2,6-dimethylpiperidine
CC1CCCC(N1)C
Flash Point 11°C (51°F)
Quantity 5g
Boiling Point 126°C to 129°C
Melting Point <-20°C
UN Number UN2733
Beilstein 79827
Formula Weight 113.2
Percent Purity 99%
Chemical Name or Material 2,6-Dimethylpiperidine, predominantly cis

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

RTECSNumber : OK5775000

TSCA : TSCA