Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethylpyrazine, 96%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
108-50-9
C6H8N2
108.144
MFCD00006148
HJFZAYHYIWGLNL-UHFFFAOYSA-N
pyrazine, 2,6-dimethyl,3,5-dimethylpyrazine,2,6-dimethylpiazine,2,6-dimethylparadiazine,2,6-dimethyl-1,4-diazine,ccris 2930,unii-n77q72c9i3,fema no. 3273,natural,2,6-dimethyl pyrazine
7938
2,6-dimethylpyrazine
CC1=CN=CC(=N1)C
108-50-9
C6H8N2
108.144
MFCD00006148
HJFZAYHYIWGLNL-UHFFFAOYSA-N
pyrazine, 2,6-dimethyl,3,5-dimethylpyrazine,2,6-dimethylpiazine,2,6-dimethylparadiazine,2,6-dimethyl-1,4-diazine,ccris 2930,unii-n77q72c9i3,fema no. 3273,natural,2,6-dimethyl pyrazine
7938
2,6-dimethylpyrazine
CC1=CN=CC(=N1)C
Flash Point 52°C
Density 52g/mL
Melting Point 34.0°C to 40.0°C
Color Yellow
Quantity 25g
Boiling Point 154.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Beilstein 23, 97
Formula Weight 108.14
Physical Form Low Melting Crystalline Solid
Percent Purity 96%
Chemical Name or Material 2, 6-Dimethylpyrazine, Balance 2, 5-isomer
Show More Show Less

GHS H Statement:
Flammable solid.
Harmful if swallowed.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning