Alfa Aesar™ 2,6-Dimethylpyrazine, 99%

Manufacturer: Alfa Aesar™ B2468706

Quantity 5g

 View more versions of this product

Catalog No. AAB2468706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 108-50-9
    Molecular Formula C6H8N2
    Formula Weight 108.14
    MDL Number MFCD00006148
    UN Number UN1325
    Beilstein 1726
    Chemical Name or Material 2,6-Dimethylpyrazine

    Specifications

    Formula Weight 108.14
    TSCA TSCA
    MDL Number MFCD00006148
    UN Number UN1325
    Health Hazard 2 GHS H Statement
    H228-H302-H315-H319-H335
    Flammable solid.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Flash Point 52°C (125°F)
    Melting Point 38° to 43°C
    Molecular Formula C6H8N2
    CAS 108-50-9
    Beilstein 1726
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P280h-P305+P351+P338
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    RTECS Number UQ2975000
    Odor Aromatic
    Boiling Point 153° to 154°C
    Quantity 5g
    Chemical Name or Material 2,6-Dimethylpyrazine
    Assay 99%

    Description

    Applications
    2,6-Dimethylpyrazine is used Preparation of chocolate, coffee, meat and nuts and other flavors

    Solubility
    Soluble in water.

    Notes
    Keep container tightly closed. Store away from oxidizing agents.

    GHS H Statement
    H228-H302-H315-H319-H335
    Flammable solid.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P280h-P305+P351+P338
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit