Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,6-Dimethylpyrazine, 99%

Click to view available options
Quantity:
25g
100g
108-50-9
C6H8N2
108.144
MFCD00006148
HJFZAYHYIWGLNL-UHFFFAOYSA-N
pyrazine, 2,6-dimethyl,3,5-dimethylpyrazine,2,6-dimethylpiazine,2,6-dimethylparadiazine,2,6-dimethyl-1,4-diazine,ccris 2930,unii-n77q72c9i3,fema no. 3273,natural,2,6-dimethyl pyrazine
7938
2,6-dimethylpyrazine
CC1=CN=CC(=N1)C
108-50-9
C6H8N2
108.144
MFCD00006148
HJFZAYHYIWGLNL-UHFFFAOYSA-N
pyrazine, 2,6-dimethyl,3,5-dimethylpyrazine,2,6-dimethylpiazine,2,6-dimethylparadiazine,2,6-dimethyl-1,4-diazine,ccris 2930,unii-n77q72c9i3,fema no. 3273,natural,2,6-dimethyl pyrazine
7938
2,6-dimethylpyrazine
CC1=CN=CC(=N1)C
Applications
2,6-Dimethylpyrazine is used Preparation of chocolate, coffee, meat and nuts and other flavors

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Odor Aromatic
Quantity 100g
Flash Point 52°C (125°F)
Boiling Point 153°C to 154°C
Melting Point 38°C to 43°C
UN Number UN1325
Beilstein 1726
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 2,6-Dimethylpyrazine

GHS H Statement
H228-H302-H315-H319-H335
Flammable solid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P280h-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : UQ2975000

TSCA : TSCA