Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dithiopurine, 94%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
5437-25-2
C5H4N4S2
184.235
MFCD00464894
VQPMXSMUUILNFZ-UHFFFAOYSA-N
2,6-dimercaptopurine,2,6-dithiopurine,dithioxanthine,9h-purine-2,6-dithiol,purine-2,6-dithiol,7h-purine-2,6-dithiol,1h-purine-2,6 3h,9h-dithione,2,6-dithioxanthine,2,6-dimercapto-7h-purine,2,6-purinedithiol
1268252
3,7-dihydropurine-2,6-dithione
C1=NC2=C(N1)C(=S)NC(=S)N2
5437-25-2
C5H4N4S2
184.235
MFCD00464894
VQPMXSMUUILNFZ-UHFFFAOYSA-N
2,6-dimercaptopurine,2,6-dithiopurine,dithioxanthine,9h-purine-2,6-dithiol,purine-2,6-dithiol,7h-purine-2,6-dithiol,1h-purine-2,6 3h,9h-dithione,2,6-dithioxanthine,2,6-dimercapto-7h-purine,2,6-purinedithiol
1268252
3,7-dihydropurine-2,6-dithione
C1=NC2=C(N1)C(=S)NC(=S)N2
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Brown
CAS Min % 92.0
CAS Max % 100.0
Assay Percent Range 94%
Formula Weight 184.25
Physical Form Powder
Percent Purity ≥92%
Chemical Name or Material 2, 6-Dithiopurine
Show More Show Less