Alfa Aesar™ 2,6-Lutidine, 98+%

Manufacturer: Alfa Aesar™ A10478AP

Quantity 500mL

 View more versions of this product

Catalog No. AAA10478AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 108-48-5
    Molecular Formula C7H9N
    Formula Weight 107.16
    MDL Number MFCD00006345
    UN Number UN1993
    Beilstein 105690
    Merck Index 14,5616
    Synonym 2,6-Dimethylpyridine
    Chemical Name or Material 2,6-Lutidine

    Specifications

    Formula Weight 107.16
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    CAS 108-48-5
    UN Number UN1993
    Boiling Point 143° to 145°C
    Odor Unpleasant
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Molecular Formula C7H9N
    MDL Number MFCD00006345
    Merck Index 14,5616
    Synonym 2,6-Dimethylpyridine
    Melting Point −7°C
    Health Hazard 1 Warning
    Quantity 500mL
    TSCA TSCA
    Beilstein 105690
    Flash Point 33°C (91°F)
    Health Hazard 2 GHS H Statement
    H226-H302-H315-H319-H335
    Flammable liquid and vapor.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number OK9700000
    Chemical Name or Material 2,6-Lutidine
    Assay ≥98%

    Description

    Applications
    2,6-Lutidine is used as a solvent in organic synthesis and as a sterically hindered mild base. It is also used as a vulcanization accelerator for dyes, resins and rubber. It also acts as a food additive.

    Solubility
    Miscible with water.

    Notes
    Incompatible with acids, acid chlorides, oxidizing agents and chloroformates.

    GHS H Statement
    H226-H302-H315-H319-H335
    Flammable liquid and vapor.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit