Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-2-(6-Methoxy-2-naphthyl)propionic Acid 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
23981-80-8
C14H14O3
230.263
MFCD00439456
CMWTZPSULFXXJA-UHFFFAOYSA-N
(+/-)-6-Methoxy-alpha-methyl-2-naphthaleneacetic Acid, (+/-)-Naproxen
1302
2-(6-methoxynaphthalen-2-yl)propanoic acid
CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O
23981-80-8
C14H14O3
230.263
MFCD00439456
CMWTZPSULFXXJA-UHFFFAOYSA-N
(+/-)-6-Methoxy-alpha-methyl-2-naphthaleneacetic Acid, (+/-)-Naproxen
1302
2-(6-methoxynaphthalen-2-yl)propanoic acid
CC(C1=CC2=C(C=C1)C=C(C=C2)OC)C(=O)O
Quantity 25g
Melting Point 157°C
Color Undesignated
UN Number 2811
Formula Weight 230.26
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material (+/-)-2-(6-Methoxy-2-naphthyl)propionic Acid