Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Pyridinedicarboxylic acid chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
3739-94-4
C7H3Cl2NO2
204.01
MFCD00006289
GWHOGODUVLQCEB-UHFFFAOYSA-N
2,6-pyridinedicarbonyl dichloride,2,6-pyridinedicarboxylic acid chloride,pyridine-2,6-dicarbonyl dichloride,2,6-pyridinedicarbonyl chloride,pyridine-2,6-dicarboxylic acid chloride,2,6-pyridinedicarboxylicacidchloride,pyridine-2,6-dicarboyl dichloride,2,6-pyridinedicarbony chloride,pubchem13243,tpc-i153
77335
pyridine-2,6-dicarbonyl chloride
C1=CC(=NC(=C1)C(=O)Cl)C(=O)Cl
3739-94-4
C7H3Cl2NO2
204.01
MFCD00006289
GWHOGODUVLQCEB-UHFFFAOYSA-N
2,6-pyridinedicarbonyl dichloride,2,6-pyridinedicarboxylic acid chloride,pyridine-2,6-dicarbonyl dichloride,2,6-pyridinedicarbonyl chloride,pyridine-2,6-dicarboxylic acid chloride,2,6-pyridinedicarboxylicacidchloride,pyridine-2,6-dicarboyl dichloride,2,6-pyridinedicarbony chloride,pubchem13243,tpc-i153
77335
pyridine-2,6-dicarbonyl chloride
C1=CC(=NC(=C1)C(=O)Cl)C(=O)Cl
Color White to Brown
Melting Point 57.0°C to 61.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 284.0°C
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (C5H3N)(COCl)2
Beilstein 22, 155
Formula Weight 204.01
Physical Form Crystalline Powder, Crystals or Chunks
Percent Purity 97%
Chemical Name or Material 2, 6-Pyridinedicarboxylic acid chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 223-125-4