Alfa Aesar™ 2,7-Di-tert-butylfluorene, 98%

Manufacturer: Alfa Aesar™ H5543006

Quantity 5g

 View more versions of this product

Catalog No. AAH5543006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 58775-05-6
    Molecular Formula C21H26
    Formula Weight 278.43
    MDL Number MFCD03093998
    Chemical Name or Material 2,7-Di-tert-butylfluorene

    Specifications

    Formula Weight 278.43
    Molecular Formula C21H26
    MDL Number MFCD03093998
    CAS 58775-05-6
    Melting Point 121° to 124°C
    Quantity 5g
    Chemical Name or Material 2,7-Di-tert-butylfluorene
    Assay 98%

    Description

    Applications
    It is used as an organic light emitting diode (OLED) intermediate.

    Solubility
    Soluble in toluene, hot methanol.

    Notes
    Store at -20°C. Avoid heat and oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit