Alfa Aesar™ 2,7-Diaminofluorene, 97+%

Manufacturer: Alfa Aesar™ H5540203

Quantity 1g

 View more versions of this product

Catalog No. AAH5540203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 525-64-4
    Molecular Formula C13H12N2
    Formula Weight 196.25
    MDL Number MFCD00001128
    Beilstein 2099859
    Merck Index 14,4156
    Synonym 2,7-Fluorenediamine
    Chemical Name or Material 2,7-Diaminofluorene

    Specifications

    Formula Weight 196.25
    CAS 525-64-4
    Merck Index 14,4156
    Synonym 2,7-Fluorenediamine
    Molecular Formula C13H12N2
    MDL Number MFCD00001128
    Beilstein 2099859
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number LL6980000
    Melting Point 160° to 162°C
    Quantity 1g
    Chemical Name or Material 2,7-Diaminofluorene
    Assay ≥97%

    Description

    Applications
    It is used as substitute for the carcinogenic benzidine in the chromogenic assay of peroxidases.

    Solubility
    Slightly soluble in cold water. Soluble in hot water. Sparingly soluble in ethanol. and acetone. Insoluble in ether.

    Notes
    Store at 4°C. Keep away from oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit