Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Diaminofluorene dihydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
13548-69-1
C13H12N2·2HCl
269.17
MFCD00012535
QJXYCUYLZDQRIN-UHFFFAOYSA-N
9h-fluorene-2,7-diamine dihydrochloride,2,7-diaminofluorene dihydrochloride,2,7-diamino-fluoren dihydrochloride,2,7-diaminofluorene di hydrochloride,2,7-diaminofluorenedihydrochloride,2,7-diaminofluorene2hcl,2,7-fluorenyldiamine2hcl,acmc-209c0q,2,7-diaminofluorene di hcl,2,7-fluorenediammonium dichloride
11543576
9H-fluorene-2,7-diamine;dihydrochloride
C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)N.Cl.Cl
13548-69-1
C13H12N2·2HCl
269.17
MFCD00012535
QJXYCUYLZDQRIN-UHFFFAOYSA-N
9h-fluorene-2,7-diamine dihydrochloride,2,7-diaminofluorene dihydrochloride,2,7-diamino-fluoren dihydrochloride,2,7-diaminofluorene di hydrochloride,2,7-diaminofluorenedihydrochloride,2,7-diaminofluorene2hcl,2,7-fluorenyldiamine2hcl,acmc-209c0q,2,7-diaminofluorene di hcl,2,7-fluorenediammonium dichloride
11543576
9H-fluorene-2,7-diamine;dihydrochloride
C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)N.Cl.Cl
Melting Point 260°C to 310°C
Color Beige-Gray to White
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Solubility Information Solubility in water: slightly soluble in water. Other solubilities: sparingly soluble in ethanol and acetone,insoluble in ether
Formula Weight 269.17
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2, 7-Diaminofluorene dihydrochloride
Show More Show Less

GHS H Statement:
Suspected of causing genetic defects.
Suspected of causing cancer.
Harmful if swallowed.

GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 236-924-8